Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sulfur trioxide pyridine complex, technical, 48-50% active SO3
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
Specifications
Specifications
| Melting Point | 155.0°C |
| Color | White to Beige |
| Infrared Spectrum | Authentic |
| Packaging | Plastic bottle |
| Linear Formula | SO3·C5H5N |
| Quantity | 25 g |
| Beilstein | 20,19 |
| Fieser | 01,1127; 02,393; 03,275; 04,473; 08,391 |
| Solubility Information | Solubility in water: reacts. Other solubilities: soluble in conc. h2so4,hcl and hclo4,soluble in dmf |
| IUPAC Name | pyridine;sulfur trioxide |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plen |
| EINECSNumber | 247-683-3 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction