Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Taurine, 99%

Catalog No. AC166540025
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle

Catalog No. AC166540025

Supplier: Thermo Scientific Chemicals 166540025

Only null left
Add to Cart

CAS: 107-35-7 | C2H7NO3S | 125.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-35-7
Molecular Formula C2H7NO3S
Molecular Weight (g/mol) 125.14
MDL Number MFCD00008197
InChI Key XOAAWQZATWQOTB-UHFFFAOYSA-N
Synonym taurine, tauphon, ethanesulfonic acid, 2-amino, l-taurine, 2-aminoethylsulfonic acid, o-due, 2-sulfoethylamine, taufon, aminoethanesulfonic acid, aminoethylsulfonic acid
PubChem CID 1123
ChEBI CHEBI:15891
IUPAC Name 2-aminoethanesulfonic acid
SMILES NCCS(O)(=O)=O

Specifications

Melting Point 300°C
Color White
Loss on Drying 0.2% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula H2NCH2CH2SO3H
Quantity 2.5 kg
Beilstein 04, 528
Merck Index 15, 9211
Solubility Information Solubility in water: 65g/L (12°C). Other solubilities: insoluble in absolute ethanol,diethyl ether,soluble in dmso
Formula Weight 125.14
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Taurine, 99%
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-483-8
RTECSNumber WX0175000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.