Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals MOPS, 99%, for biochemistry

Catalog No. AC172630250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog No. Quantity Packaging
AC172630250 25 g Plastic bottle
AC172631000 100 g Plastic bottle
AC172635000 500 g Plastic bottle
AC172630025 2.5 kg Plastic bottle
4 options

Catalog No. AC172630250

Supplier: Thermo Scientific Chemicals 172630250

Only null left
Add to Cart

CAS: 1132-61-2 | C7H15NO4S | 209.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
Synonym 3-(N-Morpholino)propanesulfonic acid
PubChem CID 70807
ChEBI CHEBI:44115
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1

Specifications

Absorbance (1 M aq. soln.) (1 cm cell) at 0nm
Absorbance Conditions (1M aq. soln.) (1 cm cell)
CAS Min % 98.5
CAS Max % 100.0
Color White
Decomposition Information >277°C
pH 3.0 to 4.5 (1% aq. soln. at 25°C)
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 99%
Infrared Spectrum Authentic
Packaging Plastic bottle
Merck Index 15, 6352
IUPAC Name 3-morpholin-4-ylpropane-1-sulfonic acid
Formula Weight 209.26
Percent Purity ≥98.5%
Grade Biochemical
Chemical Name or Material MOPS, 99%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement Emergency Overview
Causes eye, skin, and respiratory tract irritation.
NFPA
Health: 2
Flammability:
EINECSNumber 214-478-5
RTECSNumber QE9104530
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.