Learn More
tert-Butyl hydroperoxide, 70% Solution in water
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -2.8°C |
| Color | Colorless |
| Density | 0.9400g/mL |
| Boiling Point | 37.0°C (15.0 mmHg) |
| Flash Point | 43°C |
| Packaging | Plastic bottle |
| Refractive Index | 1.3860 to 1.3880 |
| Linear Formula | (CH3)3COOH |
| Quantity | 5 g |
| Beilstein | 01,IV,1616 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. Toxic in contact with skin. Causes severe skin burns and eye damage. May cause an allergic skin reaction. Fatal if inhaled. Toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye p |
| EINECSNumber | 200-915-7 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.