Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(vinyl alcohol), 98.0%-98.8% hydrolyzed, M.W. approx. 50,000-85,000

Catalog No. AC183381000
Click to view available options
Quantity:
100 g
1 kg
2.5 kg

Indicator | CAS: 9002-89-5 | (C2H4O)n | 44.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-89-5
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 44.05
MDL Number MFCD00081922
InChI Key IMROMDMJAWUWLK-UHFFFAOYSA-N
Synonym vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval
PubChem CID 11199
IUPAC Name ethenol
SMILES OC(-*)C-*

Specifications

Melting Point 230.0°C to 240.0°C
pH 5.0 to 7.0 (4% aq. soln.)
Infrared Spectrum Authentic
Linear Formula [-CH2CH(OH)-]n
Quantity 100 g
Merck Index 15,7706
Solubility Information Solubility in water: soluble in hot water.
Viscosity 9-11 mPa.s (20°C 4% aq. sol.)
Percent Purity 98%-98.8%
Chemical Name or Material Poly(vinyl alcohol)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.