Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide-d7, for NMR, 99.5 atom % D

Catalog No. AC183600100
Click to view available options
Quantity:
1 mL
5 mL
10 mL
25 mL
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 4472-41-7 | C3H7NO | 80.138 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4472-41-7
Molecular Formula C3H7NO
MDL Number MFCD00003286
InChI Key ZMXDDKWLCZADIW-YYWVXINBSA-N
Synonym n,n-dimethylformamide-d7, dmf-d7, deuterated dmf, n,n-di 2h3 methyl 2h formamide, dimethyl formamide-d7, dimethylformamide d, heptadeutero-n,n-dimethylformamide, n,n-dimethylformamide-d7 dmf-d7, formamide-1-d, n,n-di methyl-d3, n,n-dimethylformamide-d7 99.5atom%d
PubChem CID 78225

Specifications

Physical Form Liquid
Color Colorless
Refractive Index 1.428
Flash Point 58°C
Quantity 10 mL
Packaging Glass Bottle
Deuterium (D) 99.4% min.
Water 0.1% max. (Coulometric)
Density 1.0300g/mL
Linear Formula DCON(CD3)2
SMILES CN(C)C=O
IUPAC Name 1-deuterio-N,N-bis(trideuteriomethyl)formamide
Molecular Weight (g/mol) 80.138
Formula Weight 80.15
Grade NMR
Chemical Name or Material N, N-Dimethylformamide-d7, 99.5 atom % D
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May damage the unborn child.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 224-745-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.