Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zinc nitrate hexahydrate, 98%, extra pure
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 36.0°C |
| Color | White |
| Physical Form | Adhering Crystals |
| Quantity | 1 kg |
| Assay Percent Range | 98% |
| Linear Formula | Zn(NO3)2·6H2O |
| Merck Index | 14,10144 |
| Solubility Information | Solubility in water: 1800g/L (20°C). Other solubilities: freely soluble in alcohol |
| IUPAC Name | zinc;dinitrate;hexahydrate |
| Formula Weight | 297.46 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation. Harmful if swallowed. May intensify fire; oxidizer. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye pr |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction