Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zinc bromide, 99.999%, (trace metal basis), extra pure
Click to view available options
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
Specifications
Specifications
| Boiling Point | 697.0°C |
| Melting Point | 394.0°C |
| Color | White |
| Physical Form | Crystalline Powder |
| Quantity | 50 g |
| Assay Percent Range | 99.999% |
| Linear Formula | ZnBr2 |
| Fieser | 02,463; 05,762; 08,535; 09,520; 10,461; 11,600; 13,349; 15,368; 16,389 |
| Merck Index | 15, 10328 |
| Solubility Information | Solubility in water: 4470g/L (20°C). Other solubilities: soluble in ether,solutions of alkali hydroxides,1g/0.5mL 90% alcohol |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. Causes severe skin burns and eye damage. May cause an allergic skin reaction. Toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if pre |
| EINECSNumber | 231-718-4 |
| RTECSNumber | ZH1150000 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction