Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lead(IV) oxide, 97+%, extra pure
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Specifications
| Melting Point | 290°C |
| Color | Brown to Black |
| Physical Form | Powder |
| Quantity | 2.5 kg |
| Assay Percent Range | 97+% |
| Linear Formula | PbO2 |
| Fieser | 01,5331; 02,233; 03,168; 11,282 |
| Merck Index | 15, 5462 |
| Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in hcl and glacial acetic acid,insoluble in alcohol |
| IUPAC Name | dioxolead |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Very toxic to aquatic life with long lasting effects. Harmful if inhaled. May cause damage to organs through prolonged or repeated exposure. May damage the unborn child. Suspected of damaging fertility. |
| Precautionary Statement | GHS P Statement Use personal protective equipment as required. Wash face,hands and any exposed skin thoroughly after handling. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON C |
| EINECSNumber | 215-174-5 |
| RTECSNumber | OG0700000 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction