Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium(III) nitrate nonahydrate, 99%

Catalog No. AC219200010
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Plastic bottle

Catalog No. AC219200010

Supplier: Thermo Scientific Chemicals 219200010

Only null left
Add to Cart

CAS: 7789-02-8 | CrN3O9·9H2O | 400.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-02-8
MDL Number MFCD00149659
InChI Key PHFQLYPOURZARY-UHFFFAOYSA-N
Synonym chromium iii nitrate nonahydrate, unii-d2806iol1l, nitric acid, chromium 3+ salt, nonahydrate, acmc-20alg5, bis 7-chlorophenyl sulfone, cr.3no3.9h2o, ksc495a9j, chromium trinitrate, nonahydrate, chromium iii nitrate, nonahydrate, chromium 3+ nonahydrate trinitronate
PubChem CID 9887206
IUPAC Name chromium(3+);trinitrate;nonahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3]

Specifications

Melting Point 60.0°C
Color Black to Purple
pH 2 to 3 (5% aq. soln.)
Physical Form Crystalline Chunks
Quantity 1 kg
Assay Percent Range 98.5% min. (Iodometry)
Molecular Formula CrN3O9·9H2O
Linear Formula Cr(NO3)3·9H2O
Merck Index 14,2226
Solubility Information Solubility in water: 1g/L (20°C). Other solubilities: soluble in alcohol
Molecular Weight (g/mol) 400.14
Formula Weight 400.14
Percent Purity 99%
Packaging Plastic bottle
Chemical Name or Material Chromium(III) nitrate nonahydrate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if inhaled.
Causes serious eye irritation.
May intensify fire; oxidizer.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
Call a POISON CE
RTECSNumber GB6300000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.