Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
(±)-Propylene oxide, 99.5%, extra pure
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                5 mL
                            
                        
                            
                                1 L
                            
                        
                            
                                2.5 L
                            
                        
                    
                            
                                Packaging:
                            
                        
                        
                            
                                Glass bottle
                            
                        
                    Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
            Specifications
Specifications
| Melting Point | -112.0°C | 
| Density | 0.8300g/mL | 
| Boiling Point | 34.0°C | 
| Flash Point | −37°C | 
| Infrared Spectrum | Authentic | 
| Assay Percent Range | 99.4% min. (GC) | 
| Packaging | Glass bottle | 
| Refractive Index | 1.3650 to 1.3670 | 
| Quantity | 1 L | 
| UN Number | 1280 | 
| Show More | 
Safety and Handling
| Hazard Category | GHS Signal Word: Danger | 
| Hazard Statement | GHS H Statement Extremely flammable liquid and vapor. Harmful if swallowed. Toxic in contact with skin. Toxic if inhaled. May cause respiratory irritation. Causes serious eye irritation. May cause cancer.  | 
                                
| Precautionary Statement | GHS P Statement IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF SWALLOWED: rinse mouth. Do NOT induce vomi  | 
                                
| EINECSNumber | 200-879-2 | 
RUO – Research Use Only
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction