Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Petroleum ether, for analysis, boiling range 80-110°C
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
Specifications
Specifications
| Density | 0.7250g/mL |
| Boiling Point | 78°C to 113°C |
| Quantity | 1 L |
| Infrared Spectrum | Authentic |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Refractive Index | 1.3980 to 1.4060 (20°C, 589nm) |
| IUPAC Name | Petroleum ether |
| Grade | Analytical |
| Chemical Name or Material | Petroleum ether |
Safety and Handling
| Hazard Category | Danger |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction