Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Di-tert-butyl peroxide, 99%
Click to view available options
Quantity:
5 g
100 g
250 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Radical initiators
Specifications
Specifications
| Melting Point | -30°C |
| Density | 0.8000g/mL |
| Boiling Point | 109°C to 110°C |
| Flash Point | 6°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 0.1% max. Tert-butyl hydroperoxide (GC) |
| Packaging | Plastic bottle |
| Refractive Index | 1.3880 to 1.39 |
| Linear Formula | (CH3)3COOC(CH3)3 |
| Quantity | 5 g |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Heating may cause a fire. Highly flammable liquid and vapor. Suspected of causing genetic defects. Harmful to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Obtain special instructions before use. Wear protective gloves/protective clothing/eye protection/face protection. IF exposed or concerned: Get me |
| EINECSNumber | 203-733-6 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction