Learn More
3-Methyl-1-butanol, 99%, for biochemistry, AcroSeal™
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -70.0°C |
| Density | 0.8000g/mL |
| Boiling Point | 130.0°C |
| Flash Point | 45°C |
| Assay Percent Range | 98.5% min. (GC) |
| Packaging | AcroSeal™ Glass bottle |
| Refractive Index | 1.4050 to 1.4070 |
| Linear Formula | (CH3)2CHCH2CH2OH |
| Quantity | 1 L |
| Beilstein | 01,392 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Flammable liquid and vapour. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Repeated exposure may cause skin dryness or cracking. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Wash with plenty of soap and water. If skin |
| EINECSNumber | 204-633-5 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.