Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium perchlorate, ACS reagent, desiccant

Catalog No. AC413461000
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle

Catalog No. AC413461000

Supplier: Thermo Scientific Chemicals 413461000

Only null left
Add to Cart

Magnesium perchlorate, Cl2MgO8, CAS Number-10034-81-8, anhydrous magnesium perchlorate, unii-7n77z541yf, perchloric acid, magnesium salt, perchlorate de magnesium french, dehydrite, magnesium perchlorate, anhydrous, magnesium perchlorate, anhydrone, hsdb 661, magnesium diperchlorate, 500g, White | CAS: 10034-81-8 | Cl2MgO8 | 223.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent, Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-81-8
Molecular Formula Cl2MgO8
Molecular Weight (g/mol) 223.21
MDL Number MFCD00011102
InChI Key MPCRDALPQLDDFX-UHFFFAOYSA-L
Synonym anhydrone, magnesium perchlorate, dehydrite, perchloric acid, magnesium salt, anhydrous magnesium perchlorate, unii-7n77z541yf, hsdb 661, magnesium diperchlorate, magnesium perchlorate, anhydrous, perchlorate de magnesium french
PubChem CID 24840
IUPAC Name magnesium;diperchlorate
SMILES [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Mg+2]

Specifications

Melting Point 250.0°C
Color White
Physical Form Crystalline Powder, Flakes, and Chunks
Quantity 100 g
Linear Formula Mg(ClO4)2
Merck Index 15, 5742
Solubility Information Solubility in water: 993g/L (25°C). Other solubilities: very soluble in alcohol
Formula Weight 223.21
Grade ACS Reagent
Loss on Drying 8% max. (190°C)
Packaging Plastic bottle
Chemical Name or Material Magnesium perchlorate, Desiccant
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
May intensify fire; oxidizer.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Take any precaution to avoid mixing with combustibles.
IF ON SKIN: Wash
EINECSNumber 233-108-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.