Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salicylic acid, 99+%, ACS reagent

Catalog No. AC419220100
Click to view available options
Quantity:
500 g
2 kg
10 kg
Packaging:
Plastic bottle
Plastic drum

Catalog No. AC419220100

Supplier: Thermo Scientific Chemicals 419220100

Only null left
Add to Cart

CAS: 69-72-7 | C7H6O3 | 138.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-72-7
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00002439
InChI Key YGSDEFSMJLZEOE-UHFFFAOYSA-N
Synonym salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone
PubChem CID 338
ChEBI CHEBI:16914
IUPAC Name 2-hydroxybenzoic acid
SMILES OC(=O)C1=CC=CC=C1O

Specifications

Melting Point 158.0°C to 161.0°C
Boiling Point 211.0°C (20.0 mmHg)
Color White
Assay Percent Range 99+%
Beilstein 10, 43
Merck Index 15, 8469
Formula Weight 138.12
Percent Purity ≥99%
Grade ACS Reagent
Quantity 10 kg
Flash Point 157°C
Infrared Spectrum Authentic
Packaging Plastic drum
Physical Form Powder
Residue after Ignition 0.01% max.
Vapor Pressure 0.3mbar at 95°C
Chemical Name or Material Salicylic acid
Show More Show Less
Hazard Category SIGNAL WORD: DANGER!
Hazard Statement GHS P Statement Harmful if swallowed. Causes serious eye damage.
Precautionary Statement Emergency Overview
Harmful if swallowed. Irritating to eyes, respiratory system and skin.
NFPA
Health: 2
Flammability: 1
Instability: 0

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.