Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric acid, 99.6%, ACS reagent

Catalog No. AC423480020
Click to view available options
Quantity:
500 g
2 kg
Packaging:
Plastic bottle

CAS: 10043-35-3 | BH3O3 | 61.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

Color Brown-Red
Heavy Metals (as Pb) 10ppm max.
Non_volatile Matter 0.05 % max. (with MeOH)
Packaging Plastic bottle
Assay Percent Range 99.6%
Quantity 2 kg
Physical Form Crystalline powder
Linear Formula H3BO3
Fieser 01,63; 02,32; 03,29; 04,41; 05,48; 09,59; 11,70
Merck Index 15, 1339
Formula Weight 61.83
Percent Purity ≥99.5% (Acid)
Grade ACS Reagent
Chemical Name or Material Boric acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
May damage fertility. May damage the unborn child.
Precautionary Statement GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 233-139-2
RTECSNumber ED4550000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.