Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Boron trifluoride etherate, ca. 48% BF3, AcroSeal™
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
Specifications
Specifications
| Melting Point | -60.0°C |
| Color | Colorless to Straw-Yellow |
| Density | 1.1200g/mL |
| Boiling Point | 126.0°C |
| Flash Point | 58°C |
| Packaging | AcroSeal™ Glass Bottle |
| Refractive Index | 1.3429 to 1.3449 |
| Linear Formula | (C2H5)2O·BF3 |
| Quantity | 100 mL |
| Specific Gravity | 1.12 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Flammable liquid and vapor. Causes severe skin burns and eye damage. Harmful if inhaled. Causes damage to organs through prolonged or repeated exposure if inhaled. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rin |
| EINECSNumber | 203-689-8 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction