Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetaldehyde, 99.5%, extra pure, AcroSeal™

Catalog No. AC427171000
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Catalog No. Quantity Packaging
AC427171000 100 mL AcroSeal™ Glass bottle
AC427178000 800 mL AcroSeal™ Glass bottle
2 options
This item is not returnable. View return policy

Catalog No. AC427171000

Supplier: Thermo Scientific Chemicals 427171000

Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 75-07-0 | C2H4O | 44.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-07-0
Molecular Formula C2H4O
Molecular Weight (g/mol) 44.04
MDL Number MFCD00006991
InChI Key IKHGUXGNUITLKF-UHFFFAOYSA-N
Synonym ethanal, acetic aldehyde, ethyl aldehyde, acetaldehyd, acetylaldehyde, aldehyde, acetic ethanol, aldeide acetica, octowy aldehyd, aldehyde acetique
PubChem CID 177
ChEBI CHEBI:15343
IUPAC Name acetaldehyde
SMILES CC=O

Specifications

CAS Min % 99.4
CAS Max % 100.0
Melting Point -123.0°C
Color White
Density 0.7850g/mL
Boiling Point 21.0°C
Flash Point −27°C
Infrared Spectrum Authentic
Assay Percent Range 99.4% min. (GC)
Packaging AcroSeal™ Glass bottle
Linear Formula CH3CHO
Quantity 100 mL
Merck Index 15,4
Specific Gravity 0.785
Solubility Information Solubility in water: > 500g/L (20°C). Other solubilities: soluble in ethanol and acetone
Formula Weight 44.04
Percent Purity 99.5%
Grade Extra Pure
Physical Form Solid
Chemical Name or Material Acetaldehyde, Extra Pure
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye irritation.
Suspected of causing cancer.
May cause respiratory irritation.
Extremely flammable liquid and vapor.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present
EINECSNumber 200-836-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.