Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Saline-Sodium Citrate (SSC), 20X Solution (Molecular Biology), Fisher BioReagents
Shop All Fisherbrand Products

Click to view available options
Quantity:
1L
4L
20L
Description
Saline–Sodium Citrate (SSC) is used as a buffer in Southern transfer protocols.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications
Specifications
Chemical Name or Material | Saline-Sodium Citrate |
Concentration | 2.9 to 3.1 M |
pH | 7 |
Physical Form | Liquid |
Quantity | 20L |
DNase | DNase free |
Grade | Molecular Biology |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction