Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
        Fisher BioReagents™ Microbiology Media Additives: Yeast Extract
    
    
    
        
            Shop All Fisherbrand Products
        
    
    
    
    
    
    
    
    
        
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                100 g
                            
                        
                            
                                500 g
                            
                        
                            
                                2000 g
                            
                        
                    Description
Water soluble portion of autolyzed fresh yeast used in bacteriological culture media
                        
                        
                        
                        
                        
                        
                        
                        
                        
                            This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
            Specifications
Specifications
| Description | Bacterial Culture | 
| Form | Dehydrated | 
| Grade | Molecular genetics | 
| Media Type | Yeast Extract | 
| Product Type | Microbiology Media | 
| Packaging Type | Poly Bottle | 
| Quantity | 500 g | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction