Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
Agarose (Low-EEO/Multi-Purpose/Molecular Biology Grade), Fisher BioReagents™
Shop All Fisherbrand Products
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                100 g
                            
                        
                            
                                500 g
                            
                        
                    
                            
                                Packaging:
                            
                        
                        
                            
                                Poly Bottle
                            
                        
                    Description
Agarose, C12H18O9, CAS Number-9012-36-6, 500g, Gray, Cream, DNase free, 0.09 to 0.13, Pass Test, 34.5 deg. to 37.5 deg.C, 1200g/cm2min., Molecular Biology, Low-EEO/Multi-Purpose, Poly Bottle, Powder/Solid, DNase- and RNase-Free, Room temperature, 0.15% max.
 
            Specifications
Specifications
| Physical Form | Powder/Solid | 
| Color | Cream | 
| Quantity | 100 g | 
| DNase | DNase free | 
| Electroendosmosis (EEO) | 0.09 to 0.13 | 
| Gelation Temperature | 34.5° to 37.5°C | 
| Gel Strength | 1200g/cm2min. | 
| Packaging | Poly Bottle | 
| Grade | DNase- and RNase-Free | 
| Chemical Name or Material | Agarose, Low-EEO/Multi-Purpose | 
Safety and Handling
 
                        
                        | Recommended Storage | Room temperature | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
             
                                