Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eosin Y (Certified Biological Stain), Fisher Chemical™

Catalog No. E511100 Shop All Fisherbrand Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle

Catalog No. E511-100

Only null left
Add to Cart

Chemical Identifiers

CAS 17372-87-1
Molecular Formula C20H12Br4Na2O8
Molecular Weight (g/mol) 745.904
MDL Number MFCD00005040
InChI Key MASXMTNVNASWNH-UHFFFAOYSA-L
Synonym FD&C Red No. 22, Acid Red 87
PubChem CID 91886399
IUPAC Name disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate
SMILES C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]

Specifications

Bio. Stain Comm. Certification Pass Test
Dye Content 88% min.
Formula Weight 691.85
Grade Certified Biological Stain
Packaging Glass Bottle
Color Index No. 45380
Color Brown/Red
Quantity 100 g
Physical Form Solid
Chemical Name or Material Eosin Y
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.