Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrous Ammonium Sulfate Hexahydrate (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. I77212 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle

Catalog No. I77-212

Only null left
Add to Cart

Chemical Identifiers

CAS 7783-85-9
Molecular Formula FeH20N2O14S2
Molecular Weight (g/mol) 392.13
MDL Number MFCD00150530
InChI Key MQLVWQSVRZVNIP-UHFFFAOYSA-L
Synonym Mohr Salt, Iron (II) Ammonium Sulfate Hexahydrate
IUPAC Name λ²-iron(2+) diammonium hexahydrate disulfate
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Melting Point 100°C
Color Green
pH 3,5
Physical Form Solid
Quantity 2.5 kg
Assay Percent Range 98.5 to 101.5 %
Linear Formula Fe(NH4)2(SO4)2·6H2O
Formula Weight 392.14
Percent Purity 98.5 to 101.5%
Grade Certified ACS
Identification Pass test
Packaging Poly Bottle
Chemical Name or Material Ferrous Ammonium Sulfate Hexahydrate
Show More Show Less
Hazard Category Signal Word: WARNING!
Precautionary Statement Emergency Overview
Irritating to eyes, respiratory system and skin.
NFPA
Health: 1
Flammability:
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.