Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ferrous Ammonium Sulfate Hexahydrate (Crystalline/Certified ACS), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Specifications
Specifications
| Melting Point | 100°C |
| Color | Green |
| pH | 3,5 |
| Physical Form | Solid |
| Quantity | 500 g |
| Assay Percent Range | 98.5 to 101.5 % |
| Linear Formula | Fe(NH4)2(SO4)2·6H2O |
| Formula Weight | 392.14 |
| Percent Purity | 98.5 to 101.5% |
| Grade | Certified ACS |
| Show More |
Safety and Handling
| Hazard Category | Signal Word: WARNING! |
| Precautionary Statement | Emergency Overview Irritating to eyes, respiratory system and skin. NFPA Health: 1 Flammability: |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction