Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Sieves, Grade 514 (Type 4A; 8-12 Mesh Beads), Fisher Chemical
GSA_VA

Catalog No. M514500 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Color Beige
Chemical Name or Material Molecular Sieves, Grade 514
Density 2.1 g/cm3
Quantity 500 g
Mesh Size 8 to 12 Mesh
Packaging Glass Bottle
Identification Pass Test
Name Note Type 4A; 8-12 Mesh beads
Physical Form Solid
CAS 1318-02-1, 14808-60-7
Molecular Formula AlNaO6Si2
MDL Number MFCD00131613
Synonym Sodium Aluminosilicate, Aluminum Sodium Silicate
InChI Key URGAHOPLAPQHLN-UHFFFAOYSA-N
Molecular Weight (g/mol) 202.14
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.