Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Nitrate (Cryst./Certified ACS), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
500 g
3 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Specifications
Specifications
| Boiling Point | 400°C |
| Melting Point | 334°C |
| Color | White |
| pH | 6 to 8 |
| Physical Form | Solid |
| Quantity | 50 kg |
| Assay Percent Range | ≥99.0 % |
| Formula Weight | 101.1 |
| Percent Purity | ≥99.0% |
| Grade | Certified ACS |
| Show More |
Safety and Handling
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Oxidizer: Contact with combustible/organic material may cause fire. The toxicological properties have not been fully investigated. Wear personal protective equipment. Ensure adequate ventilation. Keep away from clothing and other combustible materials. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting. NFPA Health:1 Flammability:1 Instability:2 |
| DOTInformation | DOT Class 5.1, : Oxidizer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction