Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Citrate Dihydrate (Granular/Certified), Fisher Chemical™
Shop All Fisherbrand Products

Click to view available options
Quantity:
500 g
3 kg
10 kg
50 kg
275 lb.
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Decomposition Information | >230°C |
| Melting Point | 300°C |
| Color | White |
| Density | 1.67g/mL |
| pH | 7.5 to 9.5 |
| Packaging | Poly Bottle |
| Quantity | 3 kg |
| Solubility Information | 0.005% max. Insoluble Matter |
| Formula Weight | 294.1 |
| Physical Form | Solid |
| Show More |
Safety and Handling
| Hazard Statement | CAUTION! |
| Precautionary Statement | Emergency Overview May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. . NFPA Health:1 Flammability:0 Instability:0 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction