Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfate Anhydrous (Granular/Certified ACS), Fisher Chemical™

Catalog No. S4211 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
1 kg
3 kg
10 kg
50 kg
300 lb.
Packaging:
Fiber Drum
Glass Bottle
Poly Bottle
Poly Pail

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
IUPAC Name disodium sulfate
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Melting Point 884°C
Color White
pH 5 to 8
Physical Form Solid
Quantity 1 kg
Assay Percent Range ≥99.0 %
Formula Weight 142.04
Percent Purity ≥99.0%
Grade Certified ACS
Identification Pass Test
Loss on Ignition 0.5% max.
Packaging Poly Bottle
Decomposition Information Decomposition Information: >890°C
Assay ≥99.0%
Chemical Name or Material Sodium Sulfate Anhydrous
Show More Show Less
Hazard Category SIGNAL WORD: CAUTION!
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated. Hygroscopic.
NFPA
Health: 1
Flammability: 1
Instability: 0
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.