Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfate Anhydrous (Powder/USP/FCC), Fisher Chemical™

Catalog No. S42912 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
250 lb.
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
S429-12 12 kg Poly Pail
S429-500 500 g Poly Bottle
S429-212 2.5 kg Poly Bottle
S429-250LB 250 lb. Fiber Drum
4 options

Catalog No. S429-12

Only null left
Add to Cart

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
IUPAC Name disodium sulfate
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Melting Point 884°C
Color White
pH 5 to 8
Physical Form Solid
Quantity 12 kg
Formula Weight 142.04
Percent Purity 99.0 to 100.5% (FCC)
Grade FCC/USP
Identification Pass Test (FCC), Pass Test (USP), Pass Test (USP/FCC)
Loss on Drying 0.5% max. (USP), 0.5% max. (USP), 1% max. (FCC), 1% max. (FCC)
Packaging Poly Pail
Decomposition Information Decomposition Information: >890°C
Chemical Name or Material Sodium Sulfate Anhydrous
Show More Show Less
Hazard Category SIGNAL WORD: CAUTION!
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated. Hygroscopic.
NFPA
Health: 1
Flammability: 1
Instability: 0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.