Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Sulfate Anhydrous (Powder/USP/FCC), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
250 lb.
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Specifications
Specifications
| Melting Point | 884°C |
| Color | White |
| pH | 5 to 8 |
| Physical Form | Solid |
| Quantity | 250 lb. |
| Formula Weight | 142.04 |
| Percent Purity | ≥99.0% (USP) |
| Grade | FCC/USP |
| Identification | Pass Test (FCC), Pass Test (USP), Pass Test (USP/FCC) |
| Loss on Drying | 0.5% max. (USP), 0.5% max. (USP), 1% max. (FCC), 1% max. (FCC) |
| Show More |
Safety and Handling
| Hazard Category | SIGNAL WORD: CAUTION! |
| Hazard Statement | CAUTION! |
| Precautionary Statement | Emergency Overview May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated. Hygroscopic. NFPA Health: 1 Flammability: 1 Instability: 0 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction