Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silica Gel Desiccant (60-200 Mesh/Chromatographic Grade), Fisher Chemical™

Catalog No. S70425 Shop All Fisherbrand Products
Click to view available options
Quantity:
10 kg
25 kg
Packaging:
Fiber Drum

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name silanedione
SMILES O=[Si]=O

Specifications

Physical Form Solid
Color Blue
Material Silica Gel
Quantity 25 kg
Identification Pass Test
Mesh Size 60 to 200 Mesh
Packaging Fiber Drum
Grade Chromatography
Chemical Name or Material Silica Gel Desiccant, 60-200 Mesh
missing translation for 'healthHazard2' CAUTION!
missing translation for 'healthHazard3' Emergency Overview
May cause skin, eye, and respiratory tract irritation. Possible cancer hazard May cause cancer based on animal data. Wear personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.