Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KHDC1 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB164201 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB164201 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB164201 Supplier Novus Biologicals Supplier No. NBP281729AF700
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KHDC1 Polyclonal antibody specifically detects KHDC1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen KHDC1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias bA257K9.4, C6orf148, chromosome 6 open reading frame 148, KH homology domain containing 1, KH homology domain-containing protein 1, MGC10818, NDG1
Host Species Rabbit
Immunogen KHDC1 antibody was raised against a 15 amino acid peptide near the amino terminus of human KHDC1. The immunogen is located within amino acids 30 - 80 of KHDC1. Amino Acid Squence: IRRNRERNSRSGKTR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 80759
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.