Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KHSRP Rabbit anti-Human, Mouse, Rat, Clone: JE41-64, Novus Biologicals™
SDP

Catalog No. NBP275550 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP275550 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP275550 Supplier Novus Biologicals Supplier No. NBP275550

Rabbit Monoclonal Antibody

KHSRP Monoclonal specifically detects KHSRP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen KHSRP
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone JE41-64
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50-1:200
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias FBP2far upstream element-binding protein 2, FUBP2p75, FUSE binding protein 2, FUSE-binding protein 2, KH type-splicing regulatory protein, KH-type splicing regulatory protein, KSRPMGC99676
Gene Symbols KHSRP
Host Species Rabbit
Immunogen Recombinant protein within Human KHSRP aa 1-220/ 712. (SwissProt: Q92945 Human; SwissProt: Q3U0V1 Mouse; SwissProt: Q99PF5 Rat)
Molecular Weight of Antigen 73 kDa
Purification Method Protein A purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 8570.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.