Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ki 16425

Catalog No. p-7116074
Click to view available options
Quantity:
5 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

LPA1 and LPA3 receptor antagonist

Ki 16425 is an antagonist of the lysophosphatidic acid receptors LPA1 and LPA3 (Ki values are 0.25 and 0.36 μM respectively, in a GTPγS binding assay). Blocks LPA-induced dephosphorylation of Yes-associated protein (YAP) and WW domain-containing transcription regulator protein 1 (TAZ), inhibiting the Hippo signaling pathway, in HEK293A cells.

Specifications

Chemical Name or Material 3-[[[4-[4-[[[1-(2-Chlorophenyl)ethoxy]carbonyl]amino]-3-methyl-5-isoxazolyl]phenyl]methyl]thio]propanoic acid
CAS 355025-24-0
Assay Percent Range >98%
Quantity 5 mg
Molecular Formula C23H23ClN2O5S
Formula Weight Observed MW: 474.96
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.