Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ki-67, Mouse Anti-Human, RB613, Clone: B56, BD Horizon™
Mouse Monoclonal Antibody
$186.00 - $406.00
Specifications
Antigen | Ki-67 |
---|---|
Clone | B56 |
Applications | Flow Cytometry (Intracellular Staining) |
Classification | Monoclonal |
Conjugate | RB613 |
Description
Ki-67 Monoclonal antibody specifically detects Ki-67 in Human (QC Testing), Mouse (Tested in Development), Rat,Dog,Chicken (Reported) samples. It is validated for Flow Cytometry (Intracellular Staining)Specifications
Ki-67 | |
Flow Cytometry (Intracellular Staining) | |
RB613 | |
Mouse | |
Human, Mouse, Rat, Dog, Chicken | |
MKI67; Antigen identified by monoclonal antibody Ki-67; KIA | |
Human Ki-67 | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
B56 | |
Monoclonal | |
Purified | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
4288 | |
IgG1 κ | |
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title