Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Ki-67, Rabbit Monoclonal Antibody

Catalog No. p-4550778 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Liquid
Prediluted
Ready-to-Use
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RM9106RQ 12 mL Ready-to-Use
RM9106S0 100 μL Concentrated
RM9106S 1 mL Concentrated
RM9106S1 500 μL Liquid
RM9106R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. RM9106RQ Supplier Epredia™ Supplier No. RM9106RQ
Only null left
Add to Cart
Add to Cart

Ideal for Immunohistochemistry (Formalin/paraffin), Epredia™ Ki-67, Rabbit Monoclonal Antibody is part of a comprehensive antibody portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

Ki-67 is a nuclear protein, which is expressed in proliferating cells. Ki-67 is preferentially expressed during late G1-, S-, M-, and G2-phases of the cell cycle, while cells in the G0 (quiescent) phase are negative for this protein.

Host Species: Rabbit

Clone: SP6

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: C-terminus

Immunogen: A synthetic peptide derived from the human Ki-67 protein

Positive Control: Spleen, tonsil, or Breast CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Ki-67
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SP6
Conjugate Unconjugated
Host Species Rabbit
Immunogen A synthetic peptide derived from the human Ki-67 protein
Quantity 12 mL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Form Ready-to-Use
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.