Learn More
Ki-67 Rabbit Polyclonal Antibody, Epredia™
Ideal for Immunohistochemistry (Formalin/paraffin), Epredia™ Ki-67, Rabbit Polyclonal Antibody provides accurate, reproducible results.
Specifications
Antigen | Ki-67 |
---|---|
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Ki-67 is a nuclear protein, which is expressed in the proliferating cells. Ki-67 is preferentially expressed during late G1-, S-, M-, and G2-phases of the cell cycle, while cells in the G0 (quiescent) phase are negative for this protein.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human and Rat. Others not tested.
Epitope: C-terminal
Immunogen: A synthetic peptide from C-terminus of human Ki-67
Molecular Weight: 345kDa and 395kDa
Positive Control: Breast CA and lymph node
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Ki-67 | |
Polyclonal | |
Rabbit | |
Cancer and Tumor Biology | |
A synthetic peptide from C-terminus of human Ki-67 | |
Primary |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.