Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ KI-7

Catalog No. 678750
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Positive allosteric modulator of A2B receptors

KI-7 is an adenosine A2B receptor positive allosteric modulator. Potentiates the NECA (Cat. No. 1691) -induced increase in cAMP production in CHO cells expressing A2B receptors. Also potentiates the effects of NECA and the A2B receptor agonist BAY 60-6583 (Cat. No. 4472) in mediating osteoblast differentiation from MSCs in vitro.

Specifications

Chemical Name or Material α-Oxo-N-phenyl-1-(phenylmethyl)-1H-indole-3-acetamide
CAS 1489263-00-4
Quantity 50 mg
Molecular Formula C23H18N2O2
Formula Weight 354.4
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.