Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ KI-7
Click to view available options
Quantity:
10 mg
50 mg
Description
KI-7 is an adenosine A2B receptor positive allosteric modulator. Potentiates the NECA (Cat. No. 1691) -induced increase in cAMP production in CHO cells expressing A2B receptors. Also potentiates the effects of NECA and the A2B receptor agonist BAY 60-6583 (Cat. No. 4472) in mediating osteoblast differentiation from MSCs in vitro.
Specifications
Specifications
Chemical Name or Material | α-Oxo-N-phenyl-1-(phenylmethyl)-1H-indole-3-acetamide |
CAS | 1489263-00-4 |
Quantity | 10 mg |
Molecular Formula | C23H18N2O2 |
Formula Weight | 354.4 |
Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction