Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ki67/MKI67 Antibody (1297A), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB302687 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302687 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302687 Supplier Novus Biologicals Supplier No. FAB7617Y
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Ki67/MKI67 Monoclonal antibody specifically detects Ki67/MKI67 in Human samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry, KnockDown, Dual RNAscope ISH-IHC

Specifications

Antigen Ki67/MKI67
Applications Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry, KnockDown
Classification Monoclonal
Clone 1297A
Conjugate DyLight 680
Dilution Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, Knockout Validated, Dual RNAscope ISH-IHC
Formulation 50mM Sodium Borate
Gene Alias antigen identified by monoclonal Ki-67, antigen KI-67, Antigen Ki67, Ki67, Ki-67, KIA, Marker Of Proliferation Ki-67, MIB-, MIB-1, PPP1R105, Proliferation Marker Protein Ki-67, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105
Host Species Rabbit
Immunogen Human/Mouse Ki67/MKI67 synthetic peptide, Accession # E9PVX6
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Mitotic Regulators, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4288
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.