Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ki67/MKI67 Antibody (1297A), HRP, Novus Biologicals™
SDP

Catalog No. NBP254791H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP254791H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP254791H Supplier Novus Biologicals Supplier No. NBP254791H
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Ki67/MKI67 Monoclonal specifically detects Ki67/MKI67 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated.

Specifications

Antigen Ki67/MKI67
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay
Classification Monoclonal
Clone 1297A
Conjugate HRP
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated
Formulation PBS with No Preservative
Gene Alias antigen identified by monoclonal Ki-67, antigen KI-67, Antigen Ki67, Ki67, Ki-67, KIA, Marker Of Proliferation Ki-67, MIB-, MIB-1, PPP1R105, Proliferation Marker Protein Ki-67, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105
Gene Symbols MKI67
Host Species Rabbit
Immunogen The immunogen for this KI67/MKI67 Antibody (1297A) was made using a synthetic peptide from the internal portion of Mouse KI67/MKI67, between amino acids 1850-1950 [UniProt# E9PVX6].
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Mitotic Regulators, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4288
Test Specificity Ki67/MKI67 Antibody (1297A).
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.