Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ki67/MKI67 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 15 publications
Supplier: Novus Biologicals NB11090592
Description
Ki67/MKI67 Polyclonal specifically detects Ki67/MKI67 in Human, Mouse, Zebrafish samples. It is validated for Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Ki67/MKI67 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
antigen identified by monoclonal Ki-67, antigen KI-67, Antigen Ki67, Ki67, Ki-67, KIA, Marker Of Proliferation Ki-67, MIB-, MIB-1, PPP1R105, Proliferation Marker Protein Ki-67, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105 | |
Rabbit | |
359 kDa | |
0.1 mL | |
Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Mitotic Regulators, Neuroscience | |
4288 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
1 mg/mL | |
Immunohistochemistry 1:3200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:3200. Use reported in scientific literature (PMID 24599134), Immunohistochemistry-Frozen reported in scientific literature (PMID 25647012), Chromatin Immunoprecipitation (ChIP) | |
P46013 | |
MKI67 | |
The immunogen for this KI67/MKI67 Antibody was made using a synthetic peptide from the internal region of Human KI67/MKI67, between amino acids 1200-1300 [Uniprot: P46013]. | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Zebrafish | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction