Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ki67/MKI67 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 15 publications
$207.00 - $501.50
Specifications
Antigen | Ki67/MKI67 |
---|---|
Concentration | 1 mg/mL |
Dilution | Immunohistochemistry 1:3200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:3200. Use reported in scientific literature (PMID 24599134), Immunohistochemistry-Frozen reported in scientific literature (PMID 25647012), Chromatin Immunoprecipitation (ChIP) |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11090592T
![]() |
Novus Biologicals
NB11090592SS |
0.025 mL |
Each for $207.00
|
|
|||||
NB11090592
![]() |
Novus Biologicals
NB11090592 |
0.1 mL |
Each for $501.50
|
|
|||||
Description
Ki67/MKI67 Polyclonal specifically detects Ki67/MKI67 in Human, Mouse, Zebrafish samples. It is validated for Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Ki67/MKI67 | |
Immunohistochemistry 1:3200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:3200. Use reported in scientific literature (PMID 24599134), Immunohistochemistry-Frozen reported in scientific literature (PMID 25647012), Chromatin Immunoprecipitation (ChIP) | |
Polyclonal | |
Rabbit | |
Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Mitotic Regulators, Neuroscience | |
PBS with 0.02% Sodium Azide | |
antigen identified by monoclonal Ki-67, antigen KI-67, Antigen Ki67, Ki67, Ki-67, KIA, Marker Of Proliferation Ki-67, MIB-, MIB-1, PPP1R105, Proliferation Marker Protein Ki-67, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105 | |
MKI67 | |
IgG | |
Affinity Purified | |
359 kDa |
1 mg/mL | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
RUO | |
Human, Mouse, Zebrafish | |
P46013 | |
4288 | |
The immunogen for this KI67/MKI67 Antibody was made using a synthetic peptide from the internal region of Human KI67/MKI67, between amino acids 1200-1300 [Uniprot: P46013]. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title