Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ki67/MKI67 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB500170 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500170 0.1 mL
NB500170SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB500170 Supplier Novus Biologicals Supplier No. NB500170
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ki67/MKI67 Polyclonal specifically detects Ki67/MKI67 in Avian, Human, Mouse, Pig, Rat samples. It is validated for Flow Cytometry, Gene Knock-Out, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry Whole-Mount, Immunoprecipitation, Western Blot.

Specifications

Antigen Ki67/MKI67
Applications Flow Cytometry, Gene Knock-Out, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry (Whole-Mount), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, Flow Cytometry reported in scientific literature (PMID 34946963), Immunohistochemistry 1:50-1:300, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunoprecipitation reported by customer review, Immunohistochemistry-Paraffin 1:50-1:300, Immunohistochemistry-Frozen 1:50-1:300. Use reported in scientific literature (PMID 22493503), Immunoblotting reported in scientific literature (PMID 24248265), Knockout Validated
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. P46013, Q91VE6
Gene Alias antigen identified by monoclonal Ki-67, antigen KI-67, Antigen Ki67, antiKi67, Ki67, Ki-67, Ki67 ihc, Ki-67 ihc, Ki67 mouse, Ki-67 mouse, Ki67 western blot, Ki-67 western blot, KIA, Marker Of Proliferation Ki-67, MIB-, MIB-1, PPP1R105, Proliferation Marker Protein Ki-67, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105
Gene Symbols MKI67
Host Species Rabbit
Immunogen The immunogen for this KI67/MKI67 Antibody was made using a synthetic peptide from the internal region of Human KI67/MKI67, between amino acids: 1218-1232 1582-1596, 2310-2324 [Uniprot: P46013].
Molecular Weight of Antigen 359 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Mitotic Regulators, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4288
Test Specificity Human, mouse, rat (pubmed 22521325) and porcine.
Target Species Avian, Human, Mouse, Pig, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.