Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ki67/MKI67 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$268.00 - $550.50
Specifications
| Antigen | Ki67/MKI67 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1-2 ug/ml, Flow Cytometry reported in scientific literature (PMID 34946963), Immunohistochemistry 1:50-1:300, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunoprecipitation reported by customer review, Immunohistochemistry-Paraffin 1:50-1:300, Immunohistochemistry-Frozen 1:50-1:300. Use reported in scientific literature (PMID 22493503), Immunoblotting reported in scientific literature (PMID 24248265), Knockout Validated |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Immunoassay |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB500170SS
![]() |
Novus Biologicals
NB500170SS |
0.025 mL |
Each for $268.00
|
|
|||||
NB500170
![]() |
Novus Biologicals
NB500170 |
0.1 mL |
Each for $550.50
|
|
|||||
Description
Ki67/MKI67 Polyclonal specifically detects Ki67/MKI67 in Human, Mouse, Rat, Porcine, Avian samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Immunoassay.Specifications
| Ki67/MKI67 | |
| Western Blot 1-2 ug/ml, Flow Cytometry reported in scientific literature (PMID 34946963), Immunohistochemistry 1:50-1:300, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunoprecipitation reported by customer review, Immunohistochemistry-Paraffin 1:50-1:300, Immunohistochemistry-Frozen 1:50-1:300. Use reported in scientific literature (PMID 22493503), Immunoblotting reported in scientific literature (PMID 24248265), Knockout Validated | |
| Polyclonal | |
| Rabbit | |
| Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Mitotic Regulators, Neuroscience | |
| PBS with 0.05% Sodium Azide | |
| antigen identified by monoclonal Ki-67, antigen KI-67, Antigen Ki67, antiKi67, Ki67, Ki-67, Ki67 ihc, Ki-67 ihc, Ki67 mouse, Ki-67 mouse, Ki67 western blot, Ki-67 western blot, KIA, Marker Of Proliferation Ki-67, MIB-, MIB-1, PPP1R105, Proliferation Marker Protein Ki-67, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105 | |
| MKI67 | |
| IgG | |
| Affinity Purified | |
| Human, mouse, rat (pubmed 22521325) and porcine. |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Immunoassay | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Pig, Avian | |
| P46013, Q91VE6 | |
| 4288 | |
| The immunogen for this KI67/MKI67 Antibody was made using a synthetic peptide from the internal region of Human KI67/MKI67, between amino acids: 1218-1232 1582-1596, 2310-2324 [Uniprot: P46013]. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 359 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title