Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ki67/MKI67 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB354892 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB354892 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB354892 Supplier Novus Biologicals Supplier No. NB11090592MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ki67/MKI67 Polyclonal antibody specifically detects Ki67/MKI67 in Human, Mouse, Zebrafish samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), ChIP assay (ChIP)

Specifications

Antigen Ki67/MKI67
Applications Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), ChIP sequencing (ChIP-seq)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Chromatin Immunoprecipitation (ChIP)
Formulation 50mM Sodium Borate
Gene Alias antigen identified by monoclonal Ki-67, antigen KI-67, Antigen Ki67, Ki67, Ki-67, KIA, Marker Of Proliferation Ki-67, MIB-, MIB-1, PPP1R105, Proliferation Marker Protein Ki-67, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105
Host Species Rabbit
Immunogen The immunogen for this KI67/MKI67 Antibody was made using a synthetic peptide from the internal region of Human KI67/MKI67, between amino acids 1200-1300 [Uniprot: P46013].
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Mitotic Regulators, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4288
Target Species Human, Mouse, Zebrafish
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.