Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ki67/MKI67 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB360311 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360311 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360311 Supplier Novus Biologicals Supplier No. NBP305537MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ki67/MKI67 Monoclonal antibody specifically detects Ki67/MKI67 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Ki67/MKI67
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias antigen identified by monoclonal Ki-67, antigen KI-67, Antigen Ki67, Ki67, Ki-67, KIA, Marker Of Proliferation Ki-67, MIB-, MIB-1, PPP1R105, Proliferation Marker Protein Ki-67, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105
Host Species Mouse
Immunogen Recombinant construct containing amino acids 956-1322 of the mouse sequence, Accession Number: XP_006507475, expressed in and purified from E. coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Mitotic Regulators, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4288
Target Species Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.