Learn More
Ki67/MKI67 Rabbit anti-Human, Mouse, Zebrafish, DyLight 650, Polyclonal, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Ki67/MKI67 |
| Applications | Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | PerCP |
| Formulation | PBS |
| Gene Alias | antigen identified by monoclonal Ki-67, antigen KI-67, Antigen Ki67, Ki67, Ki-67, KIA, Marker Of Proliferation Ki-67, MIB-, MIB-1, PPP1R105, Proliferation Marker Protein Ki-67, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105 |
| Host Species | Rabbit |
| Immunogen | The immunogen for this KI67/MKI67 Antibody was made using a synthetic peptide from the internal region of Human KI67/MKI67, between amino acids 1200-1300 [Uniprot: P46013]. |
| Purification Method | Affinity Purified |
| Quantity | 0.1 mL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.