Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIAA0460 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501556768
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-6768 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-6768 Supplier Bethyl Laboratories, Inc Supplier No. A300031AT
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

RPRD2 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include nucleoplasm; RNA polymerase II, holoenzyme; RNA polymerase II complex binding; mRNA 3′-end processing; snRNA transcription by RNA polymerase II.

Specifications

Antigen KIAA0460
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene RPRD2
Gene Accession No. Q5VT52
Gene Alias KIAA0460
Gene Symbols RPRD2
Host Species Goat
Immunogen Between 700 and 750
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23248
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.